Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GPR39 Mouse Monoclonal Antibody [clone: 189702]
Anti-GPR39 Mouse Monoclonal Antibody [clone: 189702]
Catalog # 220020-406
Supplier:  R&D Systems
Anti-GPR39 Mouse Monoclonal Antibody [clone: 189702]
Catalog # 220020-406
Supplier:  R&D Systems
Supplier Number:  MAB3348

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    G Protein-coupled Receptor 39
  • Antigen Symbol:
    GPR39
  • Clonality:
    Monoclonal
  • Clone:
    189702
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    2863
  • Antigen Synonyms:
    G-protein coupled receptor 39|GPR39
  • Amino Acid Number:
    Met1 to Val453
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    NS0 mouse myeloma cell line transfected with human GPR39 Met1-Val453; Accession # BAG37409
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220020-406
  • Supplier no.:
    MAB3348

Specifications

About this item

Type: Primary
Antigen Symbol: GPR39
Antigen Name: G protein-coupled receptor 39
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 189702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human