Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-BRCA1 Mouse Monoclonal Antibody [clone: 440621]
Catalog # 220019-684
Supplier:  R&D Systems
Anti-BRCA1 Mouse Monoclonal Antibody [clone: 440621]
Catalog # 220019-684
Supplier:  R&D Systems
Supplier Number:  MAB22101

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Breast Cancer type 1 susceptibility protein
  • Antigen Symbol:
    BRCA1
  • Clonality:
    Monoclonal
  • Clone:
    440621
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    C-Terminal
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    672
  • Antigen Synonyms:
    BROVCA1|BRCA1/BRCA2-containing complex|subunit 1|PPP1R53|breast and ovarian cancer sususceptibility protein 1|RING finger protein 53|IRIS|protein phosphatase 1|breast cancer type 1 susceptibility protein|RNF53|PNCA4|regulatory subunit 53|breast and ovarian cancer susceptibility protein 1|PSCP|BRCAI|BRCC1
  • Amino Acid Number:
    Arg1634 to Tyr1863
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, −20 to −70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, −20 to −70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at −20 to −70 °C.
  • Immunogen:
    E. coli -derived recombinant human BRCA1 C-Terminus Arg1634-Tyr1863; Accession # P38398
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220019-684
  • Supplier no.:
    MAB22101

Specifications

About this item

Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: 440621
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human