Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PDGFA Mouse Monoclonal Antibody [clone: 184805]
Anti-PDGFA Mouse Monoclonal Antibody [clone: 184805]
Catalog # 220018-744
Supplier:  R&D Systems
Anti-PDGFA Mouse Monoclonal Antibody [clone: 184805]
Catalog # 220018-744
Supplier:  R&D Systems
Supplier Number:  MAB1055

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Platelet-Derived Growth Factor Alpha
  • Antigen Symbol:
    PDGFA
  • Clonality:
    Monoclonal
  • Clone:
    184805
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Rat
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Gene ID:
    25266
  • Antigen Synonyms:
    Platelet-derived growth factor alpha polypeptide|PDGFA|Platelet derived growth factor A chain precursor|PDGF1|Platelet-derived growth factor A chain|PlateletΓÇôdirived growth factorΓÇô1|Platelet derived growth factor alpha polypeptide|PDGF-1|Platelet-derived growth factor subunit A.|PDGFA_HUMAN|PDGF AA|PDGF A chain|PDGF subunit A|PDGF-AA
  • Amino Acid Number:
    Ser87 to Arg196
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • Immunogen:
    E. coli -derived recombinant rat PDGF‑AA Ser87-Arg196; Accession # AAB59693
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220018-744
  • Supplier no.:
    MAB1055

Specifications

About this item

Type: Primary
Antigen: PDGFA
Clonality: Monoclonal
Clone: 184805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat