Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Extracellular signal-regulated kinase 1 And Extracellular signal-regulated kinase 2
- Antigen Symbol:ERK1 + ERK2
- Clonality:Monoclonal
- Clone:269434
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Antigen Synonyms:MAP kinase 3|ERK1|MAPK 2|Mitogen-activated protein kinase 2|PRKM2|ERK2|Extracellular signal-regulated kinase 2|MAPK 3|PRKM1|MAP kinase 1|p44-MAPK|ERT1|ERK-2|Mitogen-activated protein kinase 1|MAP kinase isoform p44|ERT2|PRKM3|MAPK 1|Microtubule-associated protein 2 kinase|MAPK1|p42-MAPK|Extracellular signal-regulated kinase 1|ERK-1|MAP kinase isoform p42|Insulin-stimulated MAP2 kinase|p44-ERK1|Mitogen-activated protein kinase 3|MAP kinase 2|MAPK3
- Modification:phospho-Thr202/Tyr204, phospho-Thr185/Tyr187
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:Phosphopeptide containing ERK1 T202/Y204 site
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220018-711
- Supplier no.:MAB1018
Specifications
About this item
Simple Western
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Monoclonal
Clone: 269434
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat