Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FZD3 Rat Monoclonal Antibody [clone: 169310]
Anti-FZD3 Rat Monoclonal Antibody [clone: 169310]
Supplier:  R&D Systems
Certificates
Anti-FZD3 Rat Monoclonal Antibody [clone: 169310]
Supplier:  R&D Systems

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 220018-698
  • 220018-697
  • Antigen Symbol:
  • FZD3
  • FZD3
  • Antigen Name:
  • Frizzled 3
  • Frizzled 3
  • Antigen Synonyms:
  • Fz-3
  • Fz-3
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 169310
  • 169310
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14365
  • 14365
  • Isotype:
  • IgG2a
  • IgG2a
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3 Met1-Val157; Accession # Q61086
  • Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3 Met1-Val157; Accession # Q61086
  • Amino Acid Number:
  • Met1 to Val157
  • Met1 to Val157
  • Form:
  • Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Purification:
  • Protein A or G purified from hybridoma culture supernatant
  • Protein A or G purified from hybridoma culture supernatant
  • Storage Temperature:
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
  • The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
  • The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C

Specifications