Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MMP7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 111433]
  220018-497
Anti-MMP7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 111433]
  220018-497
 :  IC9071P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Matrix Metalloproteinase 7
  • Antigen Symbol:
    MMP7
  • Clonality:
    Monoclonal
  • Clone:
    111433
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    4316
  • Antigen Synonyms:
    PUMP-1|MPSL1|MMP-7
  • Amino Acid Number:
    Leu18 to Lys267 (Ala230del)
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human MMP‑7 Leu18-Lys267 (Ala230del); Accession # NP_002414.1
  • Purification:
    Protein A or G purified from ascites
  • Cat. No.:
    220018-497
  • Supplier no.:
    IC9071P

 

 

Type: Primary
Antigen: MMP7
Clonality: Monoclonal
Clone: 111433
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human