Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:TYRO3 protein tyrosine kinase
- Antigen Symbol:TYRO3
- Clonality:Monoclonal
- Clone:96201
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:7301
- Antigen Synonyms:Sky|etk2/tyro3|tk19-1|TK19-2|Rse|Tif|AI323366|Dtk|Brt|Etk-2
- Amino Acid Number:Ala41 to Ser428 (predicted)
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dtk Ala41-Ser428 (predicted); Accession # Q06418
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220018-012
- Supplier no.:FAB859P
Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 96201
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human