Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon gamma receptor 1
- Antigen Symbol:IFNGR1
- Clonality:Monoclonal
- Clone:92101
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:3459
- Antigen Synonyms:Interferon gamma receptor 1|IFNGR1|CD119|CDw119|IFN-gamma-R1|IFN-gamma receptor 1|Interferon-y Receptor a chain
- Amino Acid Number:Glu18 to Gly245
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human IFN-γ R1 Glu18-Gly245; Accession # P15260.1
- Purification:Protein A or G purified from ascites
- Cat. No.:220017-958
- Supplier no.:FAB673P
Specifications
About this item
Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: 92101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human