Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CD226 molecule
- Antigen Symbol:CD226
- Clonality:Monoclonal
- Clone:102511
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:10666
- Antigen Synonyms:TLiSA1|PTA1|DNAM1|CD226 antigen|Adhesion Glycoprotein|Platelet and T cell Activation Antigen 1|CD226 molecule|T lineage Specific Activation Antigen 1|DNAM 1|DNAX Accessory Molecule 1
- Amino Acid Number:Glu19 to Asn247 (predicted)
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human DNAM-1 Glu19-Asn247 (predicted); Accession # Q15762
- Purification:Protein A or G purified from ascites
- Cat. No.:220017-956
- Supplier no.:FAB666P
Type: Primary
Antigen: CD226
Clonality: Monoclonal
Clone: 102511
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human