Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NRP1 Goat Polyclonal Antibody (APC (Allophycocyanin))
Anti-NRP1 Goat Polyclonal Antibody (APC (Allophycocyanin))
  220017-878
Anti-NRP1 Goat Polyclonal Antibody (APC (Allophycocyanin))
  220017-878
 :  FAB566A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Neuropilin 1
  • Antigen Symbol:
    NRP1
  • Clonality:
    Polyclonal
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    18186
  • Antigen Synonyms:
    NP1|VEGF165R|BDCA4|NRP|CD304
  • Amino Acid Number:
    Phe22 to Ala810 (Lys811Arg), Ser829 to Asp854
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant rat Neuropilin‑1 Phe22-Ala810 (Lys811Arg), Ser829-Asp854; Accession # Q9QWJ9
  • Purification:
    Antigen Affinity-purified
  • Cat. No.:
    220017-878
  • Supplier no.:
    FAB566A

 

 

Type: Primary
Antigen: NRP1
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat