Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FLT1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 141522]
Anti-FLT1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 141522]
Catalog # 220017-799
Supplier:  R&D Systems
Anti-FLT1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 141522]
Catalog # 220017-799
Supplier:  R&D Systems
Supplier Number:  FAB4711P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    FMS-Related Tyrosine Kinase 1
  • Antigen Symbol:
    FLT1
  • Clonality:
    Monoclonal
  • Clone:
    141522
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    14254
  • Antigen Synonyms:
    FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
  • Amino Acid Number:
    Ser27 to Glu759
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant mouse VEGF R1/Flt‑1 Ser27-Glu759; Accession # P35969
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-799
  • Supplier no.:
    FAB4711P

Specifications

About this item

Type: Primary
Antigen Symbol: FLT1
Antigen Name: FMS-like tyrosine kinase 1
Antigen Synonyms: AI323757, Flt-1, VEGFR-1, VEGFR1, sFlt1
Clonality: Monoclonal
Clone: 141522
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse