Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CLEC2D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 402659]
Anti-CLEC2D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 402659]
  220017-611
Anti-CLEC2D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 402659]
  220017-611
 :  FAB3480P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    C-type lectin Domain family 2, member D
  • Antigen Symbol:
    CLEC2D
  • Clonality:
    Monoclonal
  • Clone:
    402659
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    29121
  • Antigen Synonyms:
    LLT1|CLAX|OCIL
  • Amino Acid Number:
    Ser57 to Val191
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    E. coli -derived recombinant human OCIL/CLEC2d Ser57-Val191; Accession # Q9UHP7
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-611
  • Supplier no.:
    FAB3480P

 

 

Type: Primary
Antigen: CLEC2D
Clonality: Monoclonal
Clone: 402659
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: