Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-EPHB4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 395810]
Anti-EPHB4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 395810]
  220017-538
Anti-EPHB4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 395810]
  220017-538
 :  FAB3038P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    EPH Receptor B4
  • Antigen Symbol:
    EPHB4
  • Clonality:
    Monoclonal
  • Clone:
    395810
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    2050
  • Antigen Synonyms:
    EPHB4|HTK|MYK1|TYRO11
  • Amino Acid Number:
    Ala16 to Ala539
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human EphB4 Ala16-Ala539; Accession # AAH52804
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-538
  • Supplier no.:
    FAB3038P

 

 

Type: Primary
Antigen: EPHB4
Clonality: Monoclonal
Clone: 395810
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human