Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 240742]
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 240742]
Catalog # 220017-425
Supplier:  R&D Systems
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 240742]
Catalog # 220017-425
Supplier:  R&D Systems
Supplier Number:  FAB2404P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD38 molecule
  • Antigen Symbol:
    CD38
  • Clonality:
    Monoclonal
  • Clone:
    240742
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    952
  • Antigen Synonyms:
    NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
  • Amino Acid Number:
    Met1 to Ile300
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    NS0 mouse myeloma cell line transfected with human CD38 Met1-Ile300; Accession # P28907
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-425
  • Supplier no.:
    FAB2404P

Specifications

About this item

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 240742
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: