Anti-LAG3 Goat Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 220017-409
Supplier: R&D Systems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Lymphocyte-activation gene 3
- Antigen Symbol:LAG3
- Clonality:Polyclonal
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:3902
- Antigen Synonyms:CD223
- Amino Acid Number:Leu23 to Leu450
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human LAG-3 Leu23-Leu450; Accession # P18627
- Purification:Antigen Affinity-purified
- Cat. No.:220017-409
- Supplier no.:FAB2319C
Specifications
About this item
Type: Primary
Antigen: LAG3
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human