Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGB3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 256809]
Anti-ITGB3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 256809]
Catalog # 220017-386
Supplier:  R&D Systems
Anti-ITGB3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 256809]
Catalog # 220017-386
Supplier:  R&D Systems
Supplier Number:  FAB2266P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Beta 3
  • Antigen Symbol:
    ITGB3
  • Clonality:
    Monoclonal
  • Clone:
    256809
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    3690
  • Antigen Synonyms:
    platelet membrane glycoprotein IIIa|ITGB3|platelet glycoprotein IIIa|GP3A|BDPLT2|GPIIIa|GT|integrin beta-3|CD61
  • Amino Acid Number:
    Gly27 to Asp718
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta3/CD61 Gly27-Asp718; Accession # P05106
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-386
  • Supplier no.:
    FAB2266P

Specifications

About this item

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 256809
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human