Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Carbonic Anhydrase 9
- Antigen Symbol:CA9
- Clonality:Monoclonal
- Clone:303123
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 Tests
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Gene ID:768
- Antigen Synonyms:CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
- Amino Acid Number:Pro59 to Asp414
- Storage Temperature:Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX Pro59-Asp414; Accession # Q16790
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220017-366
- Supplier no.:FAB2188P
Specifications
About this item
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 303123
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human