Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-LDLR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 472413]
Anti-LDLR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 472413]
Catalog # 220017-350
Supplier:  R&D Systems
Anti-LDLR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 472413]
Catalog # 220017-350
Supplier:  R&D Systems
Supplier Number:  FAB2148P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Low-Density Lipoprotein Receptor
  • Antigen Symbol:
    LDLR
  • Clonality:
    Monoclonal
  • Clone:
    472413
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    3949
  • Antigen Synonyms:
    FHC|FH|LDLCQ2|LDL receptor|LDL-R|Low-density lipoprotein receptor
  • Amino Acid Number:
    Ala22 to Arg788
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant human LDL R Ala22-Arg788; Accession # P01130
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-350
  • Supplier no.:
    FAB2148P

Specifications

About this item

Type: Primary
Antigen: LDLR
Clonality: Monoclonal
Clone: 472413
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human