Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-FCRL5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 307307]
  220017-338
Anti-FCRL5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 307307]
  220017-338
 :  FAB20871P

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Fc Receptor-like 5
  • Antigen Symbol:
    FCRL5
  • Clonality:
    Monoclonal
  • Clone:
    307307
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Antigen Synonyms:
    PRO820|CD307|IRTA2|BXMAS1|FCRH5|CD307e
  • Amino Acid Number:
    Gln16 to Arg844
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844; Accession # AAI01067
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-338
  • Supplier no.:
    FAB20871P

 

 

Type: Primary
Antigen Symbol: FCRL5
Antigen Name:
Antigen Synonyms: FCRL3
Clonality: Monoclonal
Clone: 307307
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human