Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-KIR2DS4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 179315]
Anti-KIR2DS4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 179315]
Supplier:  R&D Systems
Certificates
Anti-KIR2DS4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 179315]
Supplier:  R&D Systems

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 102881-530
  • 102881-528
  • Antigen Symbol:
  • KIR2DS4
  • KIR2DS4
  • Antigen Name:
  • Killer cell immunoglobulin-like Receptor two Domains short cytoplasmic tail 4
  • Killer cell immunoglobulin-like Receptor two Domains short cytoplasmic tail 4
  • Antigen Synonyms:
  • NKAT8|KKA3|NKAT-8|KIR1D|CD158I|KIR412|KIR2DS1
  • NKAT8|KKA3|NKAT-8|KIR1D|CD158I|KIR412|KIR2DS1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 179315
  • 179315
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3809
  • 3809
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • BaF3 mouse pro-B cell line transfected with human KIR2DS4; Accession # P43632
  • BaF3 mouse pro-B cell line transfected with human KIR2DS4; Accession # P43632
  • Form:
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Supplied in a saline solution containing BSA and Sodium Azide.
  • Purification:
  • Protein A or G purified from hybridoma culture supernatant
  • Protein A or G purified from hybridoma culture supernatant
  • Storage Temperature:
  • Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
  • The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.

Specifications