Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Meteorin, Glial Cell Differentiation Regulator
- Antigen Symbol:METRN
- Clonality:Monoclonal
- Clone:347504
- Conjugation:Biotin
- ELISA:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:250 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Gene ID:70083
- Antigen Synonyms:c380A1.2|glial cell dferentiation regulator|Metrn|Meteorin|MGC2601.|Meteorin precursor|METRN_HUMAN|meteorin|C16orf23
- Amino Acid Number:Gly22 to Asp291
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse Meteorin Gly22-Asp291; Accession # Q8C1Q4
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:102879-126
- Supplier no.:BAM34751
Specifications
About this item
Type: Primary
Antigen: METRN
Clonality: Monoclonal
Clone: 347504
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse