Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-INHBA Mouse Monoclonal Antibody (Biotin) [clone: 69403]
Anti-INHBA Mouse Monoclonal Antibody (Biotin) [clone: 69403]
Catalog # 220014-619
Supplier:  R&D Systems
Anti-INHBA Mouse Monoclonal Antibody (Biotin) [clone: 69403]
Catalog # 220014-619
Supplier:  R&D Systems
Supplier Number:  BAM3381

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Inhibin, Beta A
  • Antigen Symbol:
    INHBA
  • Clonality:
    Monoclonal
  • Clone:
    69403
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Form:
    Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
  • Gene ID:
    3624
  • Antigen Synonyms:
    FRP|EDF
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
  • Shipping Temperature:
    The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Chinese hamster ovary cell line CHO-derived recombinant human Activin A and recombinant human Pro-Activin A
  • Purification:
    Protein A or G purified from ascites
  • Cat. No.:
    220014-619
  • Supplier no.:
    BAM3381

Specifications

About this item

Type: Primary
Antigen: INHBA
Clonality: Monoclonal
Clone: 69403
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat