Anti-FLT3L Goat Polyclonal Antibody (Biotin)
Catalog # 220013-987
Supplier: R&D Systems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fms-related tyrosine kinase 3 ligand
- Antigen Symbol:FLT3L
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Gene ID:14256
- Antigen Synonyms:FLT3L|Fms-related tyrosine kinase 3 ligand|SL cytokine|FLT3LG|FL|Flt3 ligand
- Amino Acid Number:Gly27 to Arg188
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse Flt-3 Ligand Gly27-Arg188; Accession # P49772
- Purification:Antigen Affinity-purified
- Cat. No.:220013-987
- Supplier no.:BAF427
Specifications
About this item
Type: Primary
Antigen Symbol: FLT3L
Antigen Name: FMS-like tyrosine kinase 3 ligand
Antigen Synonyms: Flt3lg, Ly72L
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse