Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:cAMP responsive element binding protein 1
- Antigen Symbol:CREB1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Gene ID:1385
- Antigen Synonyms:CREB
- Amino Acid Number:Lys136 to Asp341
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:E. coli -derived recombinant human CREB Lys136-Asp341; Accession # P16220
- Purification:Antigen Affinity-purified
- Cat. No.:220013-686
- Supplier no.:BAF2989
Specifications
About this item
Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: