Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Chordin-like 2
- Antigen Symbol:CHRDL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:25884
- Antigen Synonyms:CHL2|Chordin related protein 2|Breast tumor novel factor 1|CRDL2_HUMAN.|Chordin like protein 2 precursor|BNF1|chordin like 2
- Amino Acid Number:Arg28 to Thr451
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2 Arg28-Thr451; Accession # NP_056239
- Purification:Antigen Affinity-purified
- Cat. No.:220010-527
- Supplier no.:AF2448
Type: Primary
Antigen: CHRDL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: