Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Nuclear Receptor Subfamily 2, Group F, Member 2
- Antigen Symbol:NR2F2
- Clonality:Monoclonal
- Clone:H7147
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Gene ID:7026
- Antigen Synonyms:SVP40|NR2F1|COUPTFB|CHTD4|TFCOUP2|ARP1|COUPTFII|NF-E3
- Storage Temperature:Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
- Immunogen:Recombinant human COUP-TF II/NR2F2
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220022-077
- Supplier no.:PP-H7147-00
Specifications
About this item
Type: Primary
Antigen: NR2F2
Clonality: Monoclonal
Clone: H7147
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human