Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Peroxisome Proliferator-Activated Receptor Gamma
- Antigen Symbol:PPARG
- Clonality:Monoclonal
- Clone:A3409A
- Conjugation:Unconjugated
- Gel Shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Gene ID:5468
- Antigen Synonyms:GLM1|PPARG1|PPARG2|PPAR gamma|NR1C3|CIMT1
- Amino Acid Number:3 to 108
- Storage Temperature:Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
- Immunogen:Recombinant human PPAR gamma/NR1C3 aa 3-108
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220022-041
- Supplier no.:PP-A3409A-00
Type: Primary
Antigen: PPARG
Clonality: Monoclonal
Clone: A3409A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human