Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Interferon alpha
- Antigen Symbol:IFNalpha
- Clonality:Monoclonal
- Clone:RMMA-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:250 µg
- Neutralisation:Yes
- Antigen Synonyms:interferon alpha 1b|interferon-alpha1|leIF D|IFN-alpha 1b|IFN-alpha-1/13|interferon alpha-1/13|interferon alpha-D
- Storage Temperature:Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
- Immunogen:Mouse Interferon alpha
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220009-325
- Supplier no.:22100-1
Type: Primary
Antigen: IFNalpha
Clonality: Monoclonal
Clone: RMMA-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse