BrightSorb® High Visibility Safety Absorbent Roll, Brady®
Catalog # 10796-196
Supplier: Brady Worldwide
Specifications
- Description:Roll
- Class Type:Chemical
- Color:Black on Yellow
- Dimensions:15W" x 150L'
- Cat. No.:10796-196
- Absorbency:20 gal.
Specifications
About this item
BrightSorb® High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste. Ideal in areas with a high risk of slips, trips and falls.
- Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
- Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
- BRIGHTSORB® High Visibility Series feature safety signage and absorbency in one
- Perforations provide cost control and reduce waste by allowing you to use only what you need
- Printed Message: Warns of slips, trips and falls
- Bright Color: Warns of presence of leaks, drips or spills even in low light