Order Entry
Puerto Rico
ContactUsLinkComponent
Barrier Spill Matting, Brady®
Barrier Spill Matting, Brady®
Catalog # 10796-126
Supplier:  Brady Worldwide
undefined
Barrier Spill Matting, Brady®
Catalog # 10796-126
Supplier:  Brady Worldwide
Supplier Number:  107682

Specifications

  • Description:
    Mats and Rugs
  • Class Type:
    Universal - All Purpose
  • Color:
    Black
  • Dimensions:
    36W" x 100L'
  • Cat. No.:
    10796-126
  • Absorbency:
    25 gal.

Specifications

About this item

Keep aisles and walkways slip free

Available in a new 100' convenient length, Barrier Spill Matting's needle-punched polypropylene fibers and non-stick backing prevents fluids from penetrating through to the floor. It can absorb water, oil, or chemicals.