ContactUsLinkComponent
Rolling Friction Car Kit
Rolling Friction Car Kit
Catalog # 470219-534
Supplier:  Wards
undefined
Rolling Friction Car Kit
Catalog # 470219-534
Supplier:  Wards
Supplier Number:  PH1120147
Specifications
  • Description:
    Rolling Friction Car Kit
  • Cat. No.:
    470219-534
  • Supplier no.:
    PH1120147
Specifications
About this item
The rolling friction car kit provides four cars for exploration into the force of friction. Each car provides a set of different material wheels (wood, fabric, soft rubber, soft silicone) with a different coefficient of friction. By setting the cars in motion with equivalent velocities, one can qualitatively demonstrate the frictional properties of rolling without slipping.
Instruction manual included.