ContactUsLinkComponent
DNA Extractor Kit, Wako
DNA Extractor Kit, Wako
Catalog # 100369-898
Supplier:  Avantor
CAS Number:  
DNA Extractor Kit, Wako
Catalog # 100369-898
Supplier:  Avantor
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    DNA Extractor Kit
  • Size:
    50 test
  • Cat. No.:
    100369-898
  • Supplier no.:
    295-50201
Specifications
About this item
DNA Extractor® Kit for DNA detection and measurement in liquids isolated from the sample proteins.
  • Used for purifying DNA from human serum
  • DNA detection and mesaurement in liquids
The extraction equipment used for purifying DNA from human serum in a single tube
Kit includes Sodium Iodide Solution 1 x 26 mL , Sodium N-Lauroyl, Sarcosinate Soln. 1 x 1.2 mL, Washing Solution (A) 1 x 42 mL, Washing Solution (B) 2 x 40 mL, Glycogen Soln. 1 x 0.1 mL.
Extraction of residual DNA in biopharmaceuticals and bacterial genomic DNA. Modified procedure uses the protease K digestion in the presence of SDS, followed by extraction with organic solvents.