Order Entry
Puerto Rico
ContactUsLinkComponent
TruLine Ammonia Electrode Replacement Module, YSI
TruLine Ammonia Electrode Replacement Module, YSI
Catalog # 10810-822
Supplier:  YSI
undefined
TruLine Ammonia Electrode Replacement Module, YSI
Catalog # 10810-822
Supplier:  YSI
Supplier Number:  400366

Specifications

  • Description:
    TruLine Ammonia Replacement Membrane Modules; Pack of 4 with 60 ml of electrode filling solution
  • Calibration:
    Slope: 54 to 59 mV/decade
  • Cat. No.:
    10810-822
  • Measurement range:
    0.02 mg/mL to 17,000 mg/L
  • pH range:
    ISA buffers sample - >pH 11 - make NH₃ gaseous
  • Interference:
    Few Volatile Amines
  • Supplier no.:
    400366

Specifications

About this item

TruLine Ammonia Replacement Membrane Modules, 4 each, includes fill solution

  • 15 electrodes for 16 different parameters
  • 4 sensor technologies - gas sensing, polymer/PVC membrane, solid state, and glass sensor
  • Integrated reference (combination/full-cell)
  • Double-junction and refillable to ensure long electrode life
  • BNC connection, connects to any instrument with BNC connectors including the YSI TruLab 1320 or the YSI MultiLab 4010-2 and 4010-3 when used with a BNC adapter
  • 3 foot cable
  • Made in the U.S.A
  • Reference/electrode fill solution, ionic strength adjustor (ISA), and small bottle of standard included with each ISE
  • Complete suite of fill solutions, ISAs, and standards available to support customers after purchase
  • 12 month warranty for solid state, glass sensor, and gas-sensing ISEs
  • 9 month warranty for ISEs with polymer/PVC membrane

There are 15 electrodes in the TruLine ISE series, with each electrode measuring the effective ion concentration of a different ionic species. This provides our customer with a wide range of ISEs to choose from. ISE parameters include: Ammonia, ammonium, bromide, cadmium, calcium, chloride, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver/sulfide, and sodium. Features replaceable/refillable membrane modules (1 replacement included), pH sensor inside.

Applications: Wastewater, aquaculture, agriculture, industrial. pH shift caused by ammonia gas is correlated to conc.

Each TruLine ISE features an integrated reference. This eliminates the need for a separate reference half-cell when measuring ion concentration and is more convenient. Each ISE can also be refilled with reference/electrode filling solution, extending the life of each ISE. The electrodes also feature a double-junction reference for optimum performance in the lab.

Certifications: GLP Compliant.
Ordering information: Used with YSI TruLab 1320; MultiLab 4010-2 or 4010-3 with BNC adapter (108132Y), sold separately. The ammonia ISE is not a true double-junction electrode, but it does contain a single-junction pH electrode inside an outer membrane that only ammonia can permeate. Cyanide and sulfide standards not included with respective kits.