Orders LinkContactUsLinkComponent
ERGOMAT® Super-Safe Ergo
ERGOMAT® Super-Safe Ergo
Supplier:   Ergomat
ERGOMAT® Super-Safe Ergo
Supplier:  Ergomat
Certificates
ERGOMAT® Super-Safe Ergo
ERGOMAT® Super-Safe Ergo
2 Options Available Below
Product Details & Documents
The ERGOMAT Super-Safe Ergo is an anti-microbial mat with gripping suction cups on the underside
Silicone and latex free. Biocera A anti-microbial agent. Available in blue and green to avoid cross contamination issues. Can be made in 2’ wide or 3’ wide runners as a standard product.
It is engineered for critical environments where liquids retained on the floor could create the potential for mats to hydroplane, causing serious injury. The premium EPDM rubber construction of this mat resists a broad range of pH exposures (2 to 13.5), and tolerates caustic cleaning protocols typical of medical, pharmaceutical and food processing facilities. It can be autoclaved in a 2’ x 3’ size.
Product Family Options
Product Information
  • Color
    Width
    Length
    Availability
    Price
  • Blue
    61 cm (2 ft.)
    91 cm (3 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $249.97
    $249.97
    Each
       $249.97
    Supplier Number: SSE0203B
  • Green
    61 cm (2 ft.)
    91 cm (3 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $237.47
    $237.47
    Each
       $237.47
    Supplier Number: SSE0203G