Narrow Mouth Reagent Bottles, with Screw Cap
Catalog # 470219-160
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Narrow Mouth Reagent Bottle, 500 mL
- Environmentally Preferable:
- Cat. No.:470219-160
- Supplier no.:220283
Specifications
About this item
These Reagent bottles are made of borosilicate glass with polopropylene screw caps with PTFE liner. They feature a no-drip pour lip and a written label area. Autoclavable.