Orion™ Green pH Combination Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Green electrodes are the first laboratory glass-bulb pH electrodes to meet all the requirements of RoHS.
- Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
- Choice of refillable or low-maintenance gel-filled models
- No disposal issues or disposal cost
- pH range is 0 to 14
- Durable epoxy body
These environmentally friendly pH combination electrodes with epoxy body and waterproof BNC connector contain no lead, mercury or other hazardous substance.
Specifications
- Catalog No:
- 97014-784
- 97014-788
- 97014-782
- Body Material:
- Epoxy
- Epoxy
- Epoxy
- pH Range:
- 0 to 14
- 0 to 14
- 0 to 14
- Operating Temperature Range:
- 0 to 90°C
- 0 to 90°C
- 0 to 90°C
- Ø:
- 12 mm (16 mm with Cap)
- 12 mm (16 mm with Cap)
- 12 mm (16 mm with Cap)