ContactUsLinkComponent
Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
  10183-850
 :  Sklar
undefined
Micro-Cut Oval Biopsy Punch, OR Grade, Sklar
  10183-850
 :  Sklar
 :  90-8948
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Ø:
    3.0 mm
  • Description:
    Micro-Cut Biopsy Punch with Locking Tip
  • Cat. No.:
    10183-850
  • Shape:
    Oval
  • Supplier no.:
    90-8948
Micro-Cut Oval Biopsy Punch Forceps are a locking, double-spring plier instrument used for obtaining uterine tissue for biopsy that features razor sharp edges.
The plier handle features a horn for thumb placement as well as a hook and two finger rest to provide maximum hand comfort and greater control of the instrument. The bite of this instrument is the unique Burke-style punch and produces a 3 x 5-mm oval tissue sample. The locking wheel at the base of the shaft has a smooth, beveled side with the rest being cross-serrated which makes the lock easy to engage. This locks the jaws and the tissue sample in place while the instrument is removed from the cervical canal, thus reducing the risk of losing the tissue sample as the instrument is withdrawn.