To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Kevorkian-Younge Biopsy Punch
- Material:Stainless steel
- Tip configuration:Rectangular Bite with 4 Lower Teeth
- Cat. No.:10198-864
- Shape:Rectangular
- Supplier no.:90-6610
Specifications
About this item
Sklar's Kevorkian-Younge Biopsy Punch is a non-ratcheted, finger ring instrument used for obtaining tissue samples in gynecological procedures
The shanks are short and angle at a box lock to a long, narrow shaft that is 10 inches long. The tip has a 3mm x 9.7mm rectangular bite with 4 lower teeth to grasp and excise the tissue for biopsy.