ContactUsLinkComponent
Acinetobacter Leeds Medium, Hardy Diagnostics
Acinetobacter Leeds Medium, Hardy Diagnostics
Catalog # 10068-018
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Acinetobacter Leeds Medium, Hardy Diagnostics
Catalog # 10068-018
Supplier:  Hardy Diagnostics
Supplier Number:  G261
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Acinetobacter Leeds Medium
  • Size:
    18 mL
  • Cat. No.:
    10068-018
  • Supplier no.:
    G261
Specifications
About this item
Leeds Acinetobacter Medium is a selective and differential medium intended for use as a plating technique to obtain a pure culture of Acinetobacter.
  • Selective components inhibit bacterial growth
  • Circular, convex, smooth and opaque colonies
  • Selective, differential medium
The selective components in Leeds Acinetobacter Medium are traditionally cefsulodin and cephradine, which inhibit the growth of non-Acinetobacter gram-negative bacteria, and vancomycin, which inhibits the growth of gram-positive bacteria
Acinetobacter spp. will produce light pink mucoid colonies with a pink to mauve color diffused into the medium. The colonies are circular, convex, smooth, and opaque with entire margins of 1 to 2 mm in diameter after 24 hours at 35°C.
The presence of fructose and sucrose and neutral red, a pH indicator, allows the differentiation of bacteria based on the products released when the bacteria use fructose and sucrose as a nutrient source. The colonies of those that release acidic products are yellow, while the colonies of those that release alkaline products are red.