To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Poly-HRP (Horseradish Peroxidase)
- Size:0.25 ml
- Protein/Peptide Name:Streptavidin
- Cat. No.:PIN200
- Supplier no.:N200
Poly-HRP Streptavidin consists of streptavidin protein that is covalently conjugated to a polymerized form of horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and the conjugated HRP polymer provides a high level of activity for detection using an appropriate substrate system. This particular Poly-HRP Streptavidin product has been used primarily in sandwich ELISA applications to improve the signal provided by typical (non-Poly) streptavidin-HRP conjugates.
Features of Poly-HRP Streptavidin:
Formulation: Conjugate in PBS containing distilled glycerol, BSA and a preservative.
Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:2,000 - 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.
Formulation: Conjugate in PBS containing distilled glycerol, BSA and a preservative.
Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:2,000 - 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.