Order Entry
United States
ContactUsLinkComponent
 
Pierce™ MS-Compatible Magnetic IP Kit, streptavidin, Thermo Scientific
Catalog # PI90408
Supplier:  Invitrogen
CAS Number:  
Pierce™ MS-Compatible Magnetic IP Kit, streptavidin, Thermo Scientific
Catalog # PI90408
Supplier:  Invitrogen
Supplier Number:  90408
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    MS-Compatible Magnetic IP Kit, streptavidin
  • Size:
    40 reactions
  • Cat. No.:
    PI90408
  • Supplier no.:
    90408

Specifications

About this item

The Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kit (Streptavidin) provides mass spectrometry-friendly reagents and an optimized protocol to enable highly effective and efficient immunoprecipitation and co-immunoprecipitation of target antigens upstream of LC-MS analysis.MS-compatible—reagents directly compatible with in-solution peptide digestion, enriched samples contain minimal detergent residuals detected using LC-MSSensitive—procedure successfully enriches low abundance proteins (low ng range)Low background—binding, wash, and elution buffers optimized to minimize enrichment of background proteinsReduced antibody contamination—antibody biotinylation reduces contamination in the eluate when compared to other IP methodsRobust—procedure and reagents have been robustly tested with numerous targets to ensure consistent enrichment of low abundant proteins (ng range) with at least 2 peptides identified per proteinThe Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads and optimized buffers that are compatible with downstream LC-MS sample preparation and analysis

After the immunoprecipitation procedure, the target-enriched elution fraction is directly compatible with in-solution tryptic digestion, eliminating the need for gel purification, detergent removal, or desalting.This kit has been rigorously validated using numerous target antigens with varying expression levels, including targets previously undetected without enrichment or by western blotting. To support IP and co-IP applications using streptavidin supports, the

Ordering information: For Research Use Only. Not for use in diagnostic procedures. Streptavidin Magnetic Beads, 1 mL

IP-MS Cell Lysis Buffer, 100 mL

IP-MS Wash Buffer A, 75 mL

IP-MS Wash Buffer B, 40 mL

IP-MS Elution Buffer, 6 mL

Store in refrigerator (2–8˚C).