Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Lysine-2HCl, [4,4,5,5-D4]
L-Lysine-2HCl, [4,4,5,5-D₄], lyophilized powder for SILAC
L-Lysine-2HCl, [4,4,5,5-D₄], lyophilized powder for SILAC
L-Lysine-2HCl, [4,4,5,5-D₄], lyophilized powder for SILAC
Catalog # PI88438
Supplier:
Invitrogen
CAS Number:
Certificates
Expand image
L-Lysine-2HCl, [4,4,5,5-D₄], lyophilized powder for SILAC
Catalog # PI88438
Supplier:
Invitrogen
Supplier Number:
88438
CAS Number:
Certificates
Specifications
Size:
500 mg
Cat. No.:
PI88438
Supplier no.:
88438
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
Product Details
Documentation