Order Entry
Puerto Rico
ContactUsLinkComponent
L-Lysine-2HCl, [¹³C₆, ¹⁵N₂], lyophilized powder for SILAC
L-Lysine-2HCl, [¹³C₆, ¹⁵N₂], lyophilized powder for SILAC
Catalog # PI88432
Supplier:  Invitrogen
CAS Number:  
L-Lysine-2HCl, [¹³C₆, ¹⁵N₂], lyophilized powder for SILAC
Catalog # PI88432
Supplier:  Invitrogen
Supplier Number:  88432
CAS Number:  

Specifications

  • Size:
    500 mg
  • Cat. No.:
    PI88432
  • Supplier no.:
    88432

Specifications

About this item

 :
Storage Temperature:Refrigerator