About this item
The Pierce™ cell culture media for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
- Sterile, Endotoxin-Free and Cell Culture-Compatible
- Flexible with Liquid Media Deficient in Both L-Lysine and L-Arginine
Allows more complete proteome coverage through multiple isotopic amino acid labeling. This is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:2...8 °C