Specifications
- Size:25 plates
- Cat. No.:PI88378
- Supplier no.:88378
Specifications
About this item
Thermo Scientific™ 96-Well Plates are manufactured using high-quality polystyrene and are high-binding with black, flat well bottoms to accommodate fluorescence assays such as the Pierce™ Quantitative Fluorometric Peptide Assay.