Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, Thermo Scientific
Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, Thermo Scientific
Catalog # PI88340
Supplier:  Invitrogen
CAS Number:  
Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, Thermo Scientific
Catalog # PI88340
Supplier:  Invitrogen
Supplier Number:  88340
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range
  • Size:
    10 ml
  • Cat. No.:
    PI88340
  • Supplier no.:
    88340

Specifications

About this item

This triple quadrupole calibration solution, extended mass range, is a mixture of 14 highly pure, ionizable components designed for both positive and negative ionization calibration of triple stage quadrupole instruments.

  • Improved mass accuracy
  • Highly validated
  • Ready to use
  • Stable - store at 2 to 8 °C for 18 months

The extended range masses improve performance of the triple quadrupole mass spectrometers.

This formulation contains 14 compounds, that are specifically optimized for use with TSQ Endura™ Series and TSQ Quantiva™ series instruments. The calibration solution is also compatible with the TSQ Quantum™ series, TSQ Discovery™ series, TSQ Quantum Ultra™ series, TSQ Quantum Access™ series, and TSQ Vantage™ series instruments.

The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

Caution: For Research Use Only. Not for use in diagnostic procedures.