Pierce™ Triple Quadrupole Calibration Solution
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive mode calibration of triple stage quadrupole instruments.
- High-Purity - Mass Spectrometry Grade Reagents in a Non-Leachable Container
- Strong Peaks - Mixture of Three Tyrosine Polymers in Methanol/Formic Acid Solution
- Stable - Store at 2 to 8 °C for More than One Year
Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers
Pierce Triple Quadrupole Calibration Solution can be used to calibrate the TSQ Quantum series, TSQ Discovery series, TSQ Quantum Access Series and TSQ Vantage Series Mass Spectrometer instruments.
The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Documents
Certificates
Search by Lot Numberghs_88325
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Description:Pierce™ Triple Quadrupole calibration solution, sufficient for 10 to 20 calibrations
- Size:10 ml
- Storage Conditions:Store between 2 and 8 ˚C.
- Cat. No.:PI88325



