Accessories for Pierce™ GTPase Enrichment Kit with GTP Probe, Thermo Scientific
Catalog # PI88315
Supplier: Invitrogen
CAS Number:
Specifications
- Description:Desthiobiotin-GTP probe
- Size:16 x 12.9 µg
- Storage Conditions:Store at -80˚C
- Cat. No.:PI88315
- Supplier no.:88315