ContactUsLinkComponent
Pierce™ Primary Neuron Isolation Kit, Thermo Scientific
Pierce™ Primary Neuron Isolation Kit, Thermo Scientific
  PI88280
 :  
undefined
Pierce™ Primary Neuron Isolation Kit, Thermo Scientific
  PI88280
 :  88280
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Primary Neuron Isolation Kit
  • Size:
    1 L
  • Cat. No.:
    PI88280
  • Supplier no.:
    88280
The Pierce™ Primary Neuron Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary neurons from embryonic cortical and hippocampal tissue from mouse and rat.
  • Procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Provides a 2-fold increase in yield compared to do-it-yourself methods
  • Greater than 95% of isolated cells remain viable in culture from Day 1 to Week 4
  • Cultured neurons contain processes indicative of complete differentiation and functionality
The Primary Neuron Isolation Kit has been designed to obtain high viability and purity of primary embryonic neurons using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains tissue-specific dissociation enzymes, media formulated specifically for neuronal cell culture, and cell culture supplements that promote the isolation and growth of functional neurons. The validated protocol and reagents produce highly pure primary neurons for either short- or long-term culture, and the kit is designed for use by both experienced and non-experienced users.
 : Upon receipt, store Product 8828X at 4?C and Product 8828Y at -20?C. Product 8828X is shipped with an ice pack. Product 8828Y is shipped on dry ice.
 : Kit includes Neuronal Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), Neuronal Culture Medium, Neuronal Culture Media Supplement, Glutamine Supplement, and Neuronal Growth Supplement. Also required (but not supplied) are fresh embryonic mouse or rat tissue sample, heat-inactivated foetal bovine serum (FBS), and poly-D-lysine treated culture dishes.
 : For Research Use Only. Not for use in diagnostic procedures.