ContactUsLinkComponent
pEF1α Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
  PI88241
 :  
pEF1α Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
  PI88241
 :  88241
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    10 µg
  • Cat. No.:
    PI88241
  • Supplier no.:
    88241
The Thermo Scientific™ pEF1α Tluc16 Vector encodes the smallest known luciferase, TurboLuc™16 (Tluc16) luciferase, as a reporter under a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in mammalian cells.Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systemsTruncated EF1α promoter for constitutive Tluc16 luciferase expressionTluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family
The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression. The Tluc16 luciferase activity can be measured using the
 : For Research Use Only. Not for use in diagnostic procedures. Store in freezer (-5 to -30˚C).